Kit, Clear with Spot 2.0mL 12x32mm 9mm Thread Large Opening Vial and 0.040in PTFE/Red Rubber Septa Lined Blue Closure. These convenience kits come with 100 vials with the closures attached. Packaged in convenient vial trays.
# de catalogue 10171-052
Fournisseur: Chemglass
Customers Who Bought This Also Bought