ContactUsLinkComponent
Cloning cylinders, 8 x 8mm, 125pcs/pack
Cloning Cylinders, Glass, Chemglass Inc.
Catalog # 10170-802
Supplier:  Chemglass
Small Business Enterprise
Cloning cylinders, 8 x 8mm, 125pcs/pack
Cloning Cylinders, Glass, Chemglass Inc.
Catalog # 10170-802
Supplier:  Chemglass
Small Business Enterprise
Specifications
  • Description:
    Cloning cylinder
  • Cat. no.:
    10170-802
  • O.D.×H:
    8×8 mm
  • Supplier No.:
    CLS-1777-02
Specifications
About this item
Clone a group or single cell by surrounding them with these glass cylinders.
  • Autoclavable or can be dry heat sterilized
The cylinders are sealed to the plate with sterile silicone high vacuum grease, such as Dow Corning 976v thereby forming an isolation well.
Delivery: Supplied in package of 125 pieces.
Customers Who Bought This Also Bought