ContactUsLinkComponent
Bottle, Anaerobic, Media, 50ml, Duran* Schott borosilicate glass, Aluminum seal, Each
Media Bottles, Anerobic, Clear, Chemglass
# de catalogue 10170-774
Fournisseur:  Chemglass
Small Business Enterprise
Bottle, Anaerobic, Media, 50ml, Duran* Schott borosilicate glass, Aluminum seal, Each
Media Bottles, Anerobic, Clear, Chemglass
# de catalogue 10170-774
Fournisseur:  Chemglass
Small Business Enterprise
Spécifications
  • Cap Type:
    Aluminium Seal
  • Capacity:
    50 ml
  • Description:
    Anaerobic Bottles
  • Cat. no.:
    10170-774
  • Supplier No.:
    CLS-4217-01
Spécifications
A propos de cet article
Anaerobic media bottles are designed for maintaining anaerobic conditions, useful for studies involving methane-producing bacteria, cell culture, drug discovery, and research and development.
  • Autoclavable
  • Manufactured from Duran® Schott borosilicate glass
Each bottle has a graduation scale indicating the approximate volumes and has a writing area for identification. Bottle is supplied with a rubber stopper and aluminum seal.
Customers Who Bought This Also Bought