Evaporator/Concentrator Manifolds, Chemglass
# de catalogue 10170-668
Fournisseur: Chemglass
Small Business Enterprise
Spécifications
- Description:48 Well Evaporator/Concentrator Manifold, Replaceable Needles
- Cat. no.:10170-668
- No. of Ports:48
- Supplier No.:OP-5020-03
Spécifications
A propos de cet article
Manifold is a low cost solution for the concentration of product in 96, 48 or 24 well blocks. Delivers equal amounts of gas to each well to gently speed solvent evaporation. Needles are constructed of stainless steel. 96 well unit has the needed pressed into the block, while the 11, 15, 24 and 48 well units have removable/replaceable needles.
- Manifold is constructed of polypropylene and the base is coated aluminum
- Needles are constructed of stainless steel
- Equipped with an adjustable flow meter for controlling gas flow into the manifold
- Inlet is a 1/8" barbed fitting
Customers Who Bought This Also Bought