PTFE-Lined Closed Caps, Chemglass
# de catalogue 10170-362
Fournisseur: Chemglass
Small Business Enterprise
Spécifications
- Color:Green
- Description:Caps with PTFE Liner
- Cat. no.:10170-362
- Thread Size:13-425
- Supplier No.:CG-895-02
Spécifications
A propos de cet article
Caps features foam backed PTFE that resists attack from virtually all chemicals at room temperature, while providing a leak-tight seal.
- Temperature tolerant
- Not autoclavable
Customers Who Bought This Also Bought