ContactUsLinkComponent
Closure Only, GPI 13-425, Green, Thermoset, Foam Back PTFE
PTFE-Lined Closed Caps, Chemglass
# de catalogue 10170-362
Fournisseur:  Chemglass
Small Business Enterprise
Closure Only, GPI 13-425, Green, Thermoset, Foam Back PTFE
PTFE-Lined Closed Caps, Chemglass
# de catalogue 10170-362
Fournisseur:  Chemglass
Small Business Enterprise
Spécifications
  • Color:
    Green
  • Description:
    Caps with PTFE Liner
  • Cat. no.:
    10170-362
  • Thread Size:
    13-425
  • Supplier No.:
    CG-895-02
Spécifications
A propos de cet article
Caps features foam backed PTFE that resists attack from virtually all chemicals at room temperature, while providing a leak-tight seal.
  • Temperature tolerant
  • Not autoclavable
Customers Who Bought This Also Bought