Order Entry
ContactUsLinkComponent
Ceramic beads
VWR® Bulk Beads
# de catalogue 10158-552
Fournisseur:  VWR International
Ceramic beads
VWR® Bulk Beads
# de catalogue 10158-552
Fournisseur:  VWR International

Spécifications

  • Ø:
    1.4 mm
  • Description:
    VWR® Ceramic Beads
  • Size:
    325 g
  • Cat. no.:
    10158-552

Spécifications

A propos de cet article

Bead beating is the most effective and verstile lysis method available.

  • Certified free of RNase and DNase
  • Designed for use on all bead mill platforms
  • Effective and versatile

VWR® offers optimal flexibility with a range of bead beating lysing matrices to support efficient homogenization of a wide range of samples.

VWR® bulk lysing matrix bead media is certified RNase and DNase free to ensure high analyte integrity. Beads are designed for use on all bead mill platforms including the VWR® Mini Bead Mill.

Certifications: RNase- and DNase-free.
Customers Who Bought This Also Bought