ContactUsLinkComponent
Hand de-crimper
Manual Hand Decrimper for Aluminum Seals and Vials, Kimble Chase
Catalog # 10158-534
Hand de-crimper
Manual Hand Decrimper for Aluminum Seals and Vials, Kimble Chase
Catalog # 10158-534
Specifications
  • Description:
    Manual Decrimper
  • Cat. no.:
    10158-534
  • Seal Size:
    11 mm
  • Supplier No.:
    69904-11
Specifications
About this item
Manual hand decrimper for aluminum seals and vials
Ideal for chromatography and general crimping needs.
Customers Who Bought This Also Bought