ContactUsLinkComponent
Hand crimper
Crimpers, DWK Life Sciences
# de catalogue 10158-528
Hand crimper
Crimpers, DWK Life Sciences
# de catalogue 10158-528
Spécifications
  • Description:
    Aluminum seal manual crimper
  • Cat. no.:
    10158-528
  • Seal Size:
    20 mm
  • Supplier No.:
    69902-20
Spécifications
A propos de cet article
Manual hand crimpers for aluminum seals and vials.
Perfect for chromatography and general crimping needs.
Customers Who Bought This Also Bought