Specifications
- Sterility:Sterile
- Type:Luer Lock Tip, Tuberculin, Low Dead Space Plunger
- Volume:1 ml
- Cat. no.:10148-330
- For Use With:Human, Laboratory and Veterinary Use
- Supplier No.:RL1
Specifications
About this item
HENKE-JECT® syringes are a high quality disposable rubber-piston syringe offering substantial savings.
- ETO gas sterilized
- Latex and pyrogen free
A lubricated synthetic rubber piston allows for a smooth draw while a positive stop helps prevent accidental pullout. Bold graduations are easily read and resist wipe off.