ContactUsLinkComponent
Easy III Step Stain
EASY III™ Rapid Differential Stains, Azer Scientific
# de catalogue 10147-834
Fournisseur:  Azer Scientific Inc.
Numéro CAS:  
EASY III™ Rapid Differential Stains, Azer Scientific
# de catalogue 10147-834
Fournisseur:  Azer Scientific Inc.
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    EASY III™ Stat Pack
  • Size:
    3 oz.
  • Supplier No.:
    ES902-3
Spécifications
A propos de cet article
EASY III™ is a three step, rapid differential stain.
  • Speciment Processing in 30 Seconds
  • Applicability In Cytology for Use in Fine Needle Aspiration
  • For Differential Staining of Peripheral Blood Smears and Bone Marrow Specimens
These stains and fixatives are routinely used for differential staining of peripheral blood smears and bone marrow specimens.
This staining procedure also has extensive applicability in cytology for use in fine needle aspiration. Yields results in less than 30 seconds with the use of one fixative and two staining solutions.
Ordering information: Available in a 3 oz. Stat Pack, 16 oz. kit, and as individual components.
Customers Who Bought This Also Bought