Spécifications
- Description:EASY III™ Stat Pack
- Size:3 oz.
- Supplier No.:ES902-3
Spécifications
A propos de cet article
EASY III™ is a three step, rapid differential stain.
- Speciment Processing in 30 Seconds
- Applicability In Cytology for Use in Fine Needle Aspiration
- For Differential Staining of Peripheral Blood Smears and Bone Marrow Specimens
These stains and fixatives are routinely used for differential staining of peripheral blood smears and bone marrow specimens.
This staining procedure also has extensive applicability in cytology for use in fine needle aspiration. Yields results in less than 30 seconds with the use of one fixative and two staining solutions.