ContactUsLinkComponent
Connecting Hose
Neoprene Burner Tubing
Catalog # 470121-742
Supplier:  Wards
Connecting Hose
Neoprene Burner Tubing
Catalog # 470121-742
Supplier:  Wards
Specifications
  • Description:
    Neoprene connecting hose, 60 cm
  • Cat. no.:
    470121-742
  • Supplier No.:
    CH100A
Specifications
About this item
Connect your burner to a fuel source easily and safely.
  • Two sizes available
  • Tight secure connection to most common burners and gas sources
This neoprene hose connects your bunsen, tirrill or any other type of burner to a fuel source.
Customers Who Bought This Also Bought