SureSTART™ Crimper and Decrimper Jaw Sets for Programmable Electronic High-Power Crimp Stations, Thermo Scientific
# de catalogue 10144-782
Fournisseur: Thermo Scientific
Spécifications
- Description:Decrimp head jaws set for crimp station, 20 mm
- Cat. no.:10144-782
- Seal Size:20 mm
- Supplier No.:ECDJ-20
Spécifications
A propos de cet article
Optimise your Thermo Scientific™ programmable electronic high-power crimp station with these crimper and decrimper sets.
Customers Who Bought This Also Bought