ContactUsLinkComponent
Electronic 13mm crimp jaw set for high power electronic crimper
SureSTART™ Crimper and Decrimper Jaw Sets for Programmable Electronic High-Power Crimp Stations, Thermo Scientific
# de catalogue 10144-776
Fournisseur:  Thermo Scientific
Electronic 13mm crimp jaw set for high power electronic crimper
SureSTART™ Crimper and Decrimper Jaw Sets for Programmable Electronic High-Power Crimp Stations, Thermo Scientific
# de catalogue 10144-776
Fournisseur:  Thermo Scientific
Spécifications
  • Description:
    Crimp head jaws set for crimp station, 13 mm
  • Cat. no.:
    10144-776
  • Seal Size:
    13 mm
  • Supplier No.:
    ECMJ-13
Spécifications
A propos de cet article
Optimise your Thermo Scientific™ programmable electronic high-power crimp station with these crimper and decrimper sets.
Customers Who Bought This Also Bought