Spécifications
- Description:Crystal Violet
- Size:3.8 L (1 gal.)
- Cat. no.:10143-260
Spécifications
A propos de cet article
Gram Stain Sets include all the necessary solutions to fully differentiate Gram positive and Gram negative specimens.
- Sets include all necessary solutions
- Enables differentiation
All Gram stain and replacement sets are packaged in a styrofoam tray and come with spout caps for ease of pouring.