Spécifications
- Description:Decolorizer Replacement Set
- Size:4 x 474 mL (16 oz.)
- Cat. no.:10143-184
Spécifications
A propos de cet article
Gram Stain Sets include all the necessary solutions to fully differentiate Gram positive and Gram negative specimens.
- Sets include all necessary solutions
- Enables differentiation
All Gram stain and replacement sets are packaged in a styrofoam tray and come with spout caps for ease of pouring.