ContactUsLinkComponent
Gram Stain Replacement Kit, Contains 4 x 16oz of 75/25 Decolorizer. This solution is used to decolorize Gram negative bacteria.
VWR® Gram Stain Sets and Reagents
# de catalogue 10143-184
Fournisseur:  VWR International
Numéro CAS:  
Gram Stain Replacement Kit, Contains 4 x 16oz of 75/25 Decolorizer. This solution is used to decolorize Gram negative bacteria.
VWR® Gram Stain Sets and Reagents
# de catalogue 10143-184
Fournisseur:  VWR International
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Decolorizer Replacement Set
  • Size:
    4 x 474 mL (16 oz.)
  • Cat. no.:
    10143-184
Spécifications
A propos de cet article
Gram Stain Sets include all the necessary solutions to fully differentiate Gram positive and Gram negative specimens.
  • Sets include all necessary solutions
  • Enables differentiation
All Gram stain and replacement sets are packaged in a styrofoam tray and come with spout caps for ease of pouring.
Customers Who Bought This Also Bought