Spécifications
- Clothing Size:4XL
- Garment application:Cleanroom,Laboratory
- Length:57"
- Color:White
- Chest Size:32"
- Supplier No.:IC264SWH4X300C
Spécifications
A propos de cet article
Tyvek® IsoClean® delivers an ideal balance of protection, durability and comfort.
- Serged seams
- Bound neck
- Raglan sleeve design
- Elastic wrists
- Zipper closure
Made using a patented flash spinning process, Tyvek® provides an inherent barrier to particles, microorganisms and non-hazardous light liquid splash. Serged seams have multiple interlocking threads that are sewn around the raw edges of garment material to create a strong, stress-resistant seam. Elastic openings at wrists and high mandarin collar with adjustable snaps allow for a secure fit. Bound neck allows for lower particle shedding. Front zipper closure allows for easy donning and doffing. Raglan sleeve design and A-line silhouette allows for greater range of motion and comfort.
Clean-processed garments offer lowest level of particle shedding within DuPont product portfolio.