Order Entry
ContactUsLinkComponent
PBS 1X, pH 7.2 (Phosphate Buffered Saline); clear colorless fluid, 500ml
Phosphate Buffered Saline (PBS), Quality Biological
# de catalogue 10128-674
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Numéro CAS:  
PBS 1X, pH 7.2 (Phosphate Buffered Saline); clear colorless fluid, 500ml
Phosphate Buffered Saline (PBS), Quality Biological
# de catalogue 10128-674
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Description:
    PBS pH 7.2
  • Size:
    500 ml
  • Sterility:
    Sterile filtered
  • Shipping conditions:
    Ambient temperature
  • Test description:
    Endotoxin and mycoplasma tested
  • Shelf life:
    24 months
  • Osmolality:
    300 ±5% mOsm/Kg H₂O
  • Shipping temperature:
    Ambient
  • Supplier No.:
    114-056-101

Spécifications

A propos de cet article

PBS (phosphate buffered saline) is a balanced salt solution used for a variety biological and cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents.

  • Endotoxin and Performance tested

PBS is also useful in immunological applications, such as Western Blotting.

Certifications: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.
Caution: Research use only.
Customers Who Bought This Also Bought