Order Entry
ContactUsLinkComponent
Hanks Balanced Salt Solution
Hanks' Balanced Salt Solution (HBSS), Quality Biological
# de catalogue 10128-666
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Numéro CAS:  
Hanks Balanced Salt Solution
Hanks' Balanced Salt Solution (HBSS), Quality Biological
# de catalogue 10128-666
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Description:
    HBSS without calcium and magnesium
  • Size:
    500 ml
  • Sterility:
    Sterile filtered
  • Shelf life:
    24 months
  • Shipping temperature:
    Ambient
  • Supplier No.:
    114-052-101

Spécifications

A propos de cet article

Hanks' Balanced Salt Solution (HBSS) provides a buffering system to maintain the physiological pH range and osmotic balance of the culture media. HBSS provides cells with a source of water and essential inorganic ions, and carbohydrate as an energy source.

HBSS without Ca and Mg can also be useful for diluting cells for counting

Formulations with calcium and magnesium support cell adhesion and are generally used as transport media, or for reagent preparation. Formulations without calcium and magnesium are required for rinsing chelators from the culture before cell dissociation.

While HBSS with Ca & Mg is useful for transporting cells or tissue samples, preparing reagents, suspension cultures, as well as to resolve clumping when clumping is a problem.

Certifications: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.
Caution: Research use only.
Customers Who Bought This Also Bought