Order Entry
ContactUsLinkComponent
Williams' Medium E
Williams Medium E, Quality Biological
# de catalogue 10128-636
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Numéro CAS:  
Williams' Medium E
Williams Medium E, Quality Biological
# de catalogue 10128-636
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Description:
    Williams medium E without L-Glutamine
  • Size:
    500 ml
  • Sterility:
    Sterile filtered
  • Shipping conditions:
    Ambient temperature
  • pH:
    7.0 ±0.2
  • Shelf Life:
    24 months
  • Supplier No.:
    112-033-101

Spécifications

A propos de cet article

Dr. Williams and his coworkers’ studies on hepatocytes in culture led to the development of Williams’ Medium E.

  • Without L-glutamine

This rich medium was used to culture the hepatocytes derived from an enrichment procedure also developed by Dr. Williams and his colleagues in 1971. The work on hepatocytes from the newborn led to attempts to culture adult liver on a long term basis. Williams’ Medium E has unique components that make it a reduced serum-supplemented medium.

Sterile Filtered
Harmonization Code: 3821.00.0000
ECCN: EAR99

It is recommended to add L-glutamine to a final concentration of 2.0mM.

Williams and his coworkers' studies on hepatocytes in culture led to the development of Williams' Medium E.

Certifications: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.
Caution: Research Use Only
Customers Who Bought This Also Bought