Order Entry
Canada
ContactUsLinkComponent
Low-EDTA TE (1X), pH 8.0, Quality Biological
Catalogue # 10128-588
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Numéro CAS:  
undefined
Low-EDTA TE (1X), pH 8.0, Quality Biological
Catalogue # 10128-588
Supplier Number:  351-324-721
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Numéro CAS:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Spécifications

  • Description:
    Low-EDTA TE (1X), pH 8.0
  • Size:
    4× 100 ml
  • Sterility:
    Sterile filtered
  • Shelf life:
    36 months
  • pH:
    8.0 ±0.1
  • Shipping temperature:
    Ambient
  • Cat. no.:
    10128-588
  • pH:
    8.0
  • Supplier No.:
    351-324-721

Spécifications

À propos de cet article

Low-EDTA 1X, pH 8.0 is molecular biology grade TE buffer used to store DNA and RNA. EDTA chelates Mg2+ and other divalent metals ions - (inhibits DNAse and RNAse to suppress DNA and RNA degradation).

  • With 0.1 mM EDTA
  • Filtered through a 0.1 µm membrane
  • DNase, RNase, and Protease tested

Tris is a buffering agent to keep the solution at a defined pH - The 'Cl' or 'HCl' indicates the pH was attained using aqueous hydrochloric acid (HCl), since Tris is naturally basic. Tris–EDTA (TE) is routinely used for suspending nucleic acid samples.

Low EDTA is recommended to re-suspend final purified DNA when DNA is used for downstream PCR.

Certifications: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.
Caution: Research use only.
Frequently Bought Together