Order Entry
ContactUsLinkComponent
Water, Ultrapure for PCR, deionized, sterile
Water, Ultrapure for PCR, deionized, sterile
# de catalogue 10128-568
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Numéro CAS:  7732-18-5
Water, Ultrapure for PCR, deionized, sterile
Water, Ultrapure for PCR, deionized, sterile
# de catalogue 10128-568
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Numéro CAS:  7732-18-5

Spécifications

  • Size:
    10 ml
  • Cat. no.:
    10128-568
  • Supplier No.:
    351-161-671EA

Spécifications

Résultats des tests de spécification

  • Water, Ultrapure for PCR, deionized, sterile:

Résultats des tests de spécification

A propos de cet article

Synonymes: Hydrogen oxide
 :Eau
Point d'ébullition:100 °C (1013 hPa)
Point de Fusion:0 °C
Densité:1 g/cm³ (4 °C)
 :Length:
 :Width:
 :Height:
 :Ø
 :Thickness:
 :Pt ébul.:|Point d'ébullition:
 :Pt ébul.: 100 °C (1013 hPa)
 :Pt Fusion:|Point de fusion:
 :Pt Fusion: 0 °C
 :Pt Eclair:|Point eclaire:
 :M.W. 18,02 g/mol
Index Merck:13,10098
Point d'ébullition:100 °C (1013 hPa)
EINECS:231-791-2
 :0
 :100
Température d'entreposage:Température ambiante
 :12,10175
 :Boiling Pt:
 :Boiling Pt: 100 °C (1013 hPa)
 :Melting Pt:
 :Melting Pt: 0 °C
 :Flash Pt:
 :Flash Pt:
 :M.W. 18.02 g/mol
 :Water
Température d'entreposage:Ambient
Customers Who Bought This Also Bought