Order Entry
ContactUsLinkComponent
Sodium chloride 5 M in aqueous solution, sterile filtered
Sodium chloride 5 M in aqueous solution, sterile filtered
# de catalogue 10128-484
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Numéro CAS:  7647-14-5
Sodium chloride 5 M in aqueous solution, sterile filtered
Sodium chloride 5 M in aqueous solution, sterile filtered
# de catalogue 10128-484
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Numéro CAS:  7647-14-5

Spécifications

  • Size:
    500 ml
  • Cat. no.:
    10128-484
  • Supplier No.:
    351-036-101

Spécifications

Résultats des tests de spécification

  • in aqueous solution:

Résultats des tests de spécification

A propos de cet article

Synonymes: Saline,Sea salt
 :Sodium chloride
 :Length:
 :Width:
 :Height:
 :Ø
 :Thickness:
 :Boiling Pt:
 :Boiling Pt: 1413 °C (1013 hPa)
 :Melting Pt:
 :Melting Pt: 801 °C
 :Flash Pt:
 :Flash Pt:
 :M.W. 58.44 g/mol
EINECS:231-598-3
 :801
 :1413
Température d'entreposage:Ambient
 :12,08742
 :14,08599
Customers Who Bought This Also Bought