Magnesium chloride 1 M in aqueous solution, sterile filtered
Catalog # 10128-476
Supplier: QUALITY BIOLOGICAL, INC.
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number: 7786-30-3
Specifications
- Size:4×100 ml
- Cat. no.:10128-476
- Supplier No.:351-033-721
Specifications
Specification Test Results
- Appearance:clear colorless liquid pass
- Dnase, Rnase and Protease:not detectable
- in aqueous solution:
Specification Test Results
About this item
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt: 1412 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 715 °C
:Flash Pt:
:Flash Pt:
:M.W. 95.21 g/mol
Merck Index:13,05684
:Magnesium chloride
:1412
:715
Storage Temperature:Ambient
EINECS:232-094-6
Customers Who Bought This Also Bought