Standard Methods Agar, CRITERION™, Hardy Diagnostics
Catalog # CA89406-066
Supplier: HARDY DIAGNOSTICS CA
CAS Number:
Specifications
- Pack type:Bucket
- Size:2 kg
- Storage temperature:–2-30°C
- Cat. no.:CA89406-066
- Supplier No.:C6982
Specifications
About this item
CRITERION™ Standard Methods Agar (Plate Count Agar) is recommended for use in determining the microbial content in dairy products, food, water and other material of sanitary importance
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free
Customers Who Bought This Also Bought