Order Entry
ContactUsLinkComponent
Blood Agar Plate
Blood Agar Plate, 5% Sheep Blood in Tryptic Soy Agar (TSA) Base, 15×100 mm plate
# de catalogue CA89405-024
Fournisseur:  HARDY DIAGNOSTICS CA
Numéro CAS:  
Blood Agar Plate
Blood Agar Plate, 5% Sheep Blood in Tryptic Soy Agar (TSA) Base, 15×100 mm plate
# de catalogue CA89405-024
Fournisseur:  HARDY DIAGNOSTICS CA
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Packaging:
    10 per package
  • Size:
    1.5×10 cm
  • Storage conditions:
    2...8 °C
  • Application:
    Clinical
  • Volume:
    19 ml
  • Supplier No.:
    A10

Spécifications

A propos de cet article

Recommended for use as general purpose growth media for the isolation, cultivation, and differentiation of a wide variety of microorganisms.

  • Optimized for superior performance
  • For the non-selective cultivation of microorganisms
  • General growth medium for bacteria, yeast, and fungi
  • Polystyrene Petri plates feature raised rings for stacking ability and prevents sliding
  • Packaged in sleeves of 10 plates, in 'breathable' cellulose bags that prevents build-up of condensation and excess moisture
  • The 100 plate size box offers superior protection during shipment and is easy to open without a cutting tool

Contains 5% sheep blood from a veterinary supervised, controlled herd of donor animals. Blood is guaranteed to be from healthy antibiotic-free animals.

Customers Who Bought This Also Bought