Pearl® Adjustable Volume Micropipettes, 8-Channel
Catalog # 10127-198
Supplier: Heathrow Scientific
Specifications
- Volume:0.5 - 50 µl
- Description:Pearl™ Adjustable volume 8-channel micro pipettor
- Cat. no.:10127-198
- Color Code:Orange
- Supplier No.:HS120215
Specifications
About this item
Pearl® adjustable volume 8-channel micro pipettors feature a sleek erogonomic design and reduced plunger pressure to help reduce the chance of RSI, as the pipettes require ²/₃rd the operation pressure of most standard pipettes.
- Volume adjustment and low-force dispensing all in one button for convenience
- Slight downward slant to tip ejector button reduces hand fatigue, incidence of repetitive strain injury (RSI)
- Isolated volume locking ring prevents accidental volume changes
- Finger hook
- 3-position volume display
- Universal tip cone
- Autoclavable
All devices feature internal metal parts for increased accuracy and calabiration stability, easy volume adjustment with locking tab to prevent accidental change, and separate tip ejector and a large digital volume display.
Each pipette is tested in accordance with standards and a calibration certificate is supplied.
Certifications: CE, UKCA.
Delivery: Supplied with one year warrenty.
Customers Who Bought This Also Bought