ContactUsLinkComponent
VWR* Flask volumetric, class A, wide mouth, 100ml, pe stopper 16, serialized
VWR® Wide Mouth Volumetric Flasks with PE Stoppers, Class A, Serialized
# de catalogue 10124-200
Fournisseur:  VWR International
VWR* Flask volumetric, class A, wide mouth, 100ml, pe stopper 16, serialized
VWR® Wide Mouth Volumetric Flasks with PE Stoppers, Class A, Serialized
# de catalogue 10124-200
Fournisseur:  VWR International
Spécifications
  • Capacity:
    100 ml
  • Cat. no.:
    10124-200
  • Stopper No.:
    16
  • Stopper Type:
    Polyethylene
  • Certification:
    Individual
Spécifications
A propos de cet article
Made from borosilicate 3.3 expansion glass.
  • Borosilicate glass construction
  • Individually serialized
  • Provided with polyethylene stopper
  • Wide-mouthed design
Flasks are serialized and supplied with polyethylene stopper.
Ordering information: Flasks come with individual certificate of compliance.
Customers Who Bought This Also Bought