ContactUsLinkComponent
VWR* Flask volumetric, class A, heavy duty, wide mouth, 25ml, PTFE stopper 13, serialized
VWR® Heavy-Duty Wide Mouth Volumetric Flasks with PTFE Stoppers, Class A, Serialized
# de catalogue 10124-184
Fournisseur:  VWR International
VWR* Flask volumetric, class A, heavy duty, wide mouth, 25ml, PTFE stopper 13, serialized
VWR® Heavy-Duty Wide Mouth Volumetric Flasks with PTFE Stoppers, Class A, Serialized
# de catalogue 10124-184
Fournisseur:  VWR International
Spécifications
  • Capacity:
    25 ml
  • Cat. no.:
    10124-184
  • Stopper No.:
    13
  • Stopper Type:
    PTFE
  • Certification:
    Individual
Spécifications
A propos de cet article
Heavy-duty wide-mouth volumetric flasks are made to the highest standards to qualify for Class A certification.
  • Borosilicate glass fabrication
  • Flasks are individually serialized
Made from borosilicate 3.3 expansion glass. Flasks are serialized and supplied with PTFE stopper.
Ordering information: Flasks come with individual certificate of compliance.
Customers Who Bought This Also Bought