VWR® Color-Coded Autoclavable Instrument Marking Tape
Catalogue # 10124-144
Fournisseur: VWR International
Spécifications
- Color:Green
- Description:VWR® Color-Coded Autoclavable Instrument Marking Tape
- Dimensions:9 mm x 5 m
- Cat. no.:10124-144
Spécifications
À propos de cet article
These 9 mm x5m, vinyl, self-stick tapes color code autoclave items.
- Non-toxic
- Vinyl
- Self-stick tapes
Tapes are designed for use on items such as surgical instruments, laboratory glassware, medical waste, and more for easy identification. These tapes attach securely and safely to any clean surface and resist water, oil, and acids.
Frequently Bought Together