VWR® Color-Coded Autoclavable Instrument Marking Tape
# de catalogue 10124-142
Fournisseur: VWR International
Spécifications
- Dimensions:9 mm x 5 m
- Color:Yellow
- Cat. no.:10124-142
Spécifications
A propos de cet article
These 9 mm x5m, vinyl, self-stick tapes color code autoclave items.
- Non-toxic
- Vinyl
- Self-stick tapes
Tapes are designed for use on items such as surgical instruments, laboratory glassware, medical waste, and more for easy identification. These tapes attach securely and safely to any clean surface and resist water, oil, and acids.
Customers Who Bought This Also Bought