VWR® Heavy-Duty Wide Mouth Volumetric Flasks with Glass Stoppers, Amber Glass, Class A, Serialized
# de catalogue 10124-094
Fournisseur: VWR International
Spécifications
- Capacity:50 ml
- Cat. no.:10124-094
- Stopper No.:13
- Stopper Type:Glass pennyhead
- Certification:Individual
Spécifications
A propos de cet article
Heavy-duty wide-mouth volumetric flasks are made to the highest standards to qualify for Class A certification.
- Borosilicate glass fabrication
- Flasks are individually serialized
Made from borosilicate 3.3 expansion glass. Flasks are serialized and supplied with glass penny head stopper.
Ordering information: Flasks come with individual certificate of compliance.
Customers Who Bought This Also Bought