VWR® Heavy-Duty Wide Mouth Volumetric Flasks with Glass Stoppers, Class A, Serialized
Catalog # 10124-072
Supplier: VWR International
Specifications
- Capacity:100 ml
- Cat. no.:10124-072
- Stopper Type:Glass pennyhead
- Stopper No.:16
- Certification:Individual
Specifications
About this item
Heavy-duty wide-mouth volumetric flasks are made to the highest standards to qualify for Class A certification.
- Borosilicate glass fabrication
- Flasks are individually serialized
Made from borosilicate 3.3 expansion glass. Flasks are serialized and supplied with glass penny head stopper.
Ordering information: Flasks come with individual certificate of compliance.
Customers Who Bought This Also Bought