VWR® Volumetric Flasks without Stopper, Amber Glass, Class A, Serialized
Catalog # 10124-028
Supplier: VWR International
Specifications
- Capacity:10 ml
- Cat. no.:10124-028
- Stopper No.:9
- Certification:Individual
Specifications
About this item
Made from borosilicate 3.3 expansion glass, flasks are serialized and supplied without stopper.
- Borosilicate glass construction
- Individually serialized
- Comes without a stopper
Flasks are made with a graduation mark for accurate measurements.
Ordering information: Flasks come with individual certificate of compliance.
Customers Who Bought This Also Bought