ContactUsLinkComponent
Flux
Lithium Borate Fusion Fluxes, XRF Scientific
# de catalogue 10119-384
Fournisseur:  Kitco Metals
Lithium Borate Fusion Fluxes, XRF Scientific
# de catalogue 10119-384
Fournisseur:  Kitco Metals
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Color:
    White
  • Description:
    100% Lithium Tetraborate
  • Size:
    1 kg
  • Supplier No.:
    KSC1000-02
Spécifications
A propos de cet article
Lithium borate fusion fluxes are designed for sample preparation in XRF, AA, or ICP analysis
These prefused, granular fluxes offer the highest purities. They exhibit excellent homogeneity, and are free flowing, non-dusty, and high density. Low loss of ignition and consistent chemistry for the best results.
Fluxes are ideally suited for sample preparation for Iron Ore, Bauxite, Cement, Silica, Rocks, Ferro Alloys, Sulphides, Carbonates, and Coal Ash.
Customers Who Bought This Also Bought