Spécifications
- Color:White
- Description:100% Lithium Tetraborate
- Size:1 kg
- Supplier No.:KSC1000-02
Spécifications
A propos de cet article
Lithium borate fusion fluxes are designed for sample preparation in XRF, AA, or ICP analysis
These prefused, granular fluxes offer the highest purities. They exhibit excellent homogeneity, and are free flowing, non-dusty, and high density. Low loss of ignition and consistent chemistry for the best results.
Fluxes are ideally suited for sample preparation for Iron Ore, Bauxite, Cement, Silica, Rocks, Ferro Alloys, Sulphides, Carbonates, and Coal Ash.