Order Entry
ContactUsLinkComponent
Vial
Cardboard Storage Boxes, Globe Scientific
# de catalogue 10118-800
Fournisseur:  Globe Scientific
Vial
Cardboard Storage Boxes, Globe Scientific
# de catalogue 10118-800
Fournisseur:  Globe Scientific

Spécifications

  • Color:
    White
  • Description:
    100-Place (10 x 10) Storage Boxes
  • Cat. no.:
    10118-800
  • For Use With:
    Up to 3" x 12 mm Tubes
  • Supplier No.:
    3096

Spécifications

A propos de cet article

These cardboard storage boxes are designed for the compact storage of sample tubes, cryogenic vials, and centrifuge tubes in a temperature range of –196 to 121°C.

  • Liquid-resistant coating
  • Designed for use in refrigerators and cryogenic freezer systems
  • Drainage holes eliminate excess liquid and reduce moisture build-up
  • Numbered grid on the inside bottom of each box aids in sample identification
Ordering information: Assorted colors are available for special order.
Delivery: Supplied complete with dividers.
Customers Who Bought This Also Bought