Media Bottles, PETG, WHEATON®
# de catalogue 10112-172
Fournisseur: DWK Life Sciences
Spécifications
- Cap Type:Tamper Evident Cap
- Capacity:500 ml
- Sterility:Sterile
- Width:73 mm
- Color:Clear
- Description:Media bottles, square
- Height:172 mm
- Material:PETG
- Cat. no.:10112-172
- Packaging:12
- Cap Color:Blue
- Supplier No.:WPBGC0500S
- Height with Cap:177.7 mm
Spécifications
A propos de cet article
These bottles are made of eastman tritan eastar copolyester PETG resin for single use labware and medical devices.
- Ideal for the storage of media, laboratory reagents and biological buffers
- Graduation markings are molded in for reference filling
- Have a standard cap
- Comprised of resins that are free of animal-derived components
- Suitable for transport and storage, leak resistant
- Recommended temperature range +70 to –40 °C
The PETG is an amorphous material that displays performance properties including clarity and toughness as well as excellent gas barrier and chemical inertness. The products can be safely sterilized with gamma irradiation without property loss or color shift. They are provided gamma irradiated sterilized to SAL 10-6 with blue caps attached.
Certifications: Meets ISO 10993 and / or USP Class VI biocompatibility requirements and is Food Contact Status compliant.
Customers Who Bought This Also Bought