SVL Caps, Ace Glass Incorporated
# de catalogue 10099-446
Fournisseur: Ace Glass
Small Business Enterprise
Spécifications
- Description:Cap, Vent with Insert
- Cat. no.:10099-446
- Thread Size:SVL22
- Supplier No.:7647-40
Spécifications
A propos de cet article
Solid black or vented replacement caps with SVL thread for rotary evaporator components.
Customers Who Bought This Also Bought