SVL Caps, Ace Glass Incorporated
# de catalogue 10099-444
Fournisseur: Ace Glass
Small Business Enterprise
Spécifications
- Description:Cap, Solid
- Cat. no.:10099-444
- Thread Size:SVL30
- Supplier No.:7647-30
Spécifications
A propos de cet article
Solid black or vented replacement caps with SVL thread for rotary evaporator components.
Customers Who Bought This Also Bought