Septa for GL Caps, Silicone and Silicone/PTFE, Ace Glass Incorporated
# de catalogue 10099-412
Fournisseur: Ace Glass
Small Business Enterprise
Spécifications
- Description:Septum for GL45 cap
- Material:Silicone/PTFE
- Cat. no.:10099-412
- Supplier No.:7625-10
Spécifications
A propos de cet article
These septa are designed to fit the DURAN® open-top cap product family.
Available in both silicone and PTFE-faced silicone for good chemical resistance. Caps can withstand 130 °C (266 °F) vapor or 200 °C (392 °F) dry heat.
Customers Who Bought This Also Bought