PTFE Stopper with Extraction Nut, Ace Glass Incorporated
# de catalogue 10099-190
Fournisseur: Ace Glass
Small Business Enterprise
Spécifications
- Description:PTFE Stopper with Polypropylene Extraction Nut, Solid
- Cat. no.:10099-190
- Joint Size:34/45
- Supplier No.:12636-22
Spécifications
A propos de cet article
Stoppers are available in solid PTFE with polypropylene extraction nut.
Customers Who Bought This Also Bought