Replacement Sleeves for Laboratory Clamps, Ace Glass
Catalog # 10099-120
Supplier: Ace Glass
Small Business Enterprise
Specifications
- Description:Labjaws Replacement Sleeve Set, 2 Prong, Vinyl
- Size:Medium
- Cat. no.:10099-120
- Supplier No.:11074-03
Specifications
About this item
Red vinyl or fiberglass (high temperature) replacement sleeves are available in three grip sizes for two or three prongs
Small fits grips less than 5 cm, medium fits grips 5 to 7 cm, and large fits grips above 7 cm. Fiberglass sleeves are recommended for temperatures above 100 °C.
Customers Who Bought This Also Bought