Accessories for Virtuoso™ Vial Identification System, Thermo Scientific
# de catalogue 10081-052
Fournisseur: Thermo Scientific
Spécifications
- Description:Virtuoso crimp top, vial, clear glass, V-patch, 2 ml capacity, 11 mm cap size
- Cat. no.:10081-052
- Supplier No.:60180-VT302