To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Description:Petroleum Ether
- Size:1 L
- Chemical/Physical Specifications:Solution
- Cat. no.:470301-952
- Grade:Reagent
- Supplier No.:PP0105-1LR
- Disposal Method:This disposal method is published for your convenience. You MUST have checked with your federal, state, and local regulations before using this method and these methods are only applicable for small laboratory sized quantities.
Use a lab apron, chemical resistant gloves and goggles before following the below method to dispose of hazardous waste.
1. Place a shallow pan in the fume hood.
2. Pour the liquid to be disposed of into the pan and turn the fume hood on.
3. Over a few hours, the liquid will evaporate.
a. Be careful as these compounds are volatile, so any flames and ignition sources need to be removed.
b. If a fume hood is not available, the evaporation can take place outside.
Spécifications
A propos de cet article
CAS Number: 8032-32-4
Formula Weight: Mixture
Formula: Mixture
Hazard Info: Flammable
Density (g/mL): 060-075
Boiling Point (°C): 20-75
Freezing Point (°C): <73
Solubility: Miscible with Organic Solvents
Synonyms: Ligroin
Shelf Life (months): 12
Storage: Red
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.