Copper(II) Sulfate Pentahydrate
Catalogue # 470300-852
Fournisseur: Ward's Science
Numéro CAS:
Spécifications
- Description:Copper(II) Sulfate 5-Hydrate
- Size:500 g
- Chemical/Physical Specifications:Crystal
- Cat. no.:470300-852
- Grade:Laboratory
- Supplier No.:CC0545-500G
Spécifications
À propos de cet article
CAS Number: 7758-99-8
Formula: CuSO4 5H2O
Density: 2.284 g/mL
Freezing Point: 150°C
Solubility: Water and Methanol
Synonyms: Copper II Sulfate 5-Hydrate, Cupric Sulfate 5-Hydrate
Shelf Life: 36 Months
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Caution: This product is a toxic irritant.
Frequently Bought Together