BASIK* 1.5 ml Screw top vial and cap slit eppendorf clear
# de catalogue 10063-406
Fournisseur: MicroSolv Technology Corp
BASIK* 1.5 ml Screw top vial and cap slit eppendorf clear
# de catalogue 10063-406
Fournisseur: MicroSolv Technology Corp
Customers Who Bought This Also Bought
Chloroform ≥99.8% stabilized ACS, VWR Chemicals BDH®
Fournisseur: VWR International
558,88 $ to 2 230,71 $
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Fournisseur: MilliporeSigma
635,20 $ to 2 541,50 $
Hexane (mixture of isomers) ≥98.5% ACS for HPLC, Supelco®
Fournisseur: MilliporeSigma
649,44 $ to 2 496,86 $
Glycerine ≥99.7%, Laboratory Reagent, VWR Chemicals BDH®
Fournisseur: VWR International
881,80 $ to 2 807,56 $