ContactUsLinkComponent
Cell Scraper 39cm
VWR® Cell Scrapers
# de catalogue 10062-910
Fournisseur:  VWR International
Cell Scraper 39cm
VWR® Cell Scrapers
# de catalogue 10062-910
Fournisseur:  VWR International
Spécifications
  • Description:
    Cell Scraper for Dishes, Lifter Blade Position
  • Blade Width:
    30 mm
  • Overall Length:
    390 mm
Spécifications
A propos de cet article
Specifically designed to make collecting of cells easier and more effective.
  • Cross-ribbed handle in ABS provides a greater rigidity and ensures a better control while scraping cells
  • Thin, flexible TPE blade prevents damage to cells
  • Individually wrapped
  • Sterilized by electron beam irradiation
  • Non pyrogenic
Available with a choice of blade positions; scraper for use in flasks, or lifter for use in harvesting cells (especially stem cells) in dishes.
Customers Who Bought This Also Bought