VWR® Cell Scrapers
# de catalogue 10062-906
Fournisseur: VWR International
Spécifications
- Description:Cell Scraper for Dishes, Lifter Blade Position
- Cat. no.:10062-906
- Blade Width:20 mm
- Overall Length:250 mm
Spécifications
A propos de cet article
Specifically designed to make collecting of cells easier and more effective.
- Cross-ribbed handle in ABS provides a greater rigidity and ensures a better control while scraping cells
- Thin, flexible TPE blade prevents damage to cells
- Individually wrapped
- Sterilized by electron beam irradiation
- Non pyrogenic
Available with a choice of blade positions; scraper for use in flasks, or lifter for use in harvesting cells (especially stem cells) in dishes.
Customers Who Bought This Also Bought