GC Snap Ring Cap With Septum, 11 mm, Restek
# de catalogue 10058-676
Fournisseur: Restek
Spécifications
- Septum:PTFE/Silicone
- Color:Clear
- Description:Snap Ring Cap w/Septum
- Material:PP
- Supplier No.:21733
- Orifice:11 mm
Spécifications
A propos de cet article
Restek snap ring caps are an extremely sturdy, affordable option when crimp-top caps are not needed.
- Economical
- Sturdy polypropylene construction
Use these snap caps when you crimping is unnecessary. Snap caps are available with either liners or septa, and they seal an 11-millimeter opening.
Customers Who Bought This Also Bought