Order Entry
ContactUsLinkComponent
Tr-blue septa 9mm 50pk
Septa, Thermo Scientific
# de catalogue 10057-440
Fournisseur:  Thermo Scientific
Septa, Thermo Scientific
# de catalogue 10057-440
Fournisseur:  Thermo Scientific

Spécifications

  • Ø:
    9 mm
  • Maximum Temperature:
    200...250 °C
  • Description:
    TR-blue septa
  • Material:
    Silicone
  • Packaging:
  • Supplier No.:
    313B3240

Spécifications

A propos de cet article

Thermo Scientific™ BTO and TR-Green septa for GC instruments.

  • Variety of materials and temperature limits for all applications
  • For use in Thermo Scientific™ TRACE™ and FOCUS™ GCs and instruments from other manufacturers
  • Contaminant-free in individual blister packs

Made of low-bleed silicone, these septa have excellent mechanical properties, are ideal for demanding GC-MS applications, and may be used reliably up to 400 °C.

Customers Who Bought This Also Bought