ContactUsLinkComponent
Open-Top Screw Cap fits 7mL screw cap septum vials, 15mm closure size
Open Top Screw Caps, Thermo Scientific
# de catalogue 10056-148
Fournisseur:  Thermo Scientific
Open-Top Screw Cap fits 7mL screw cap septum vials, 15mm closure size
Open Top Screw Caps, Thermo Scientific
# de catalogue 10056-148
Fournisseur:  Thermo Scientific
Spécifications
  • Cap Size:
    15 mm-425
  • Description:
    Open-Top Screw Caps
  • Cat. no.:
    10056-148
  • Supplier No.:
    TS-13216
Spécifications
A propos de cet article
Provide inert, air-tight seal and direct puncture-access to sample with a syringe needle.
Customers Who Bought This Also Bought